Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC II HLA-DQ3 Monoclonal Antibody (2HB6)
GREENER_CHOICE

Catalog No. PIMA127170
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA127170 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA127170 Supplier Invitrogen™ Supplier No. MA127170
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-27170 detects HLA DQ3 from mouse samples.

Human Leukocyte Antigens are highly polymorphic proteins that are involved in the presentation of antigens to the T cell receptor. There are two classes of HLA antigens, class I (HLA A, HLA B and HLA C) and class II (HLA D). HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC II HLA-DQ3
Applications ELISA, Flow Cytometry, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 2HB6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias MHC DQ3
Host Species Mouse
Immunogen ATL mouse spleen cells and PRIESS (HLA-DR4 ) cells.
Purification Method IgG fraction
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.