Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC II Monomorphic Monoclonal Antibody (YKIX334.2)
GREENER_CHOICE

Catalog No. PIMA528495
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528495 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528495 Supplier Invitrogen™ Supplier No. MA528495
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Rat anti Dog MHC Class II Monomorphic antibody, clone YKIX334.2 recognizes a monomorphic epitope on canine MHC Class II and was classified at the First Canine Leucocyte Antigen Workshop (Cobbold et al. 1994).

MHC II Monomorphic is part of major histocompatibility complex (MHC) is a cluster of genes that are important in the immune response to infections. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC II Monomorphic
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone YKIX334.2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene HLA-DRB1
Gene Accession No. P18470
Gene Alias beta 1 domain; B-L; B-L beta chain; B-L beta minor; BLA-DRB3; B-LB; BLB major; BLB1; BLB2; B-LB2; B-LB21; B-LB21 major; BLB3; Bl-Beta II protein; B-LBII; BoLA-DQA; BoLA-DQA1; BOLA-DQA2; BOLA-DQA2 protein; BOLA-DQA3; BOLA-DQA5; BOLA-DQB; BOLA-DRA; BoLA-DR-alpha; BoLA-DRB; BOLA-DRB3; BoLA-DRB3 protein; BoLA-DRB3.2; Bota-DRB01; Bota-DRB02; Bota-DRB04; Bota-DRB07; Bota-DRB21; Bota-DRB22; bovine leukocyte antigen; class II histocompatibility antigen, B-L beta chain; DLA class II histocompatibility antigen, DR-1 beta chain; DLA DRBB1 beta chain; DLA-DQA; DLA-DQA1; DLA-DQB; DLA-DQB1; DLA-DR beta; DLA-DRB1; DLA-DRBB1; dog leucocyte antigen DQA; DQA; dqa 7; dqa1; DQALPHA; DQB; dqb1; DR beta-chain antigen binding domain; DRA; DRB; DRB1; DRB3; DRBB1; DR-beta chain; DR-beta chain MHC class II; ELA-DQA; ela-DRA; Eqca-DQA1; Eqca-DQA2; Eqca-DQA3; Eqca-DQB1; Eqca-DQB2; Eqca-DRA; Eqca-DRB1; Eqca-DRB2; Eqca-DRB3; GSP-BLB1; GSP-BLB2; histocompatibility complex, class II, DR beta 3; HLA-DRB1; integral membrane glycoprotein; integral membrane protein; LA-DRB; leukocyte antigen DRB3; major histocompatability complex class II; major histocompatibility class II antigen B-L beta; major histocompatibility complex; major histocompatibility complex class II B; major histocompatibility complex class II beta chain; Major histocompatibility complex class II beta chain BLB1; Major histocompatibility complex class II beta chain BLB1, (similar to HLA class II, D beta chain); major histocompatibility complex class II beta chain BLB1, (similar to HLA class II, D beta chain); Major histocompatibility complex class II beta chain BLB2, (similar to HLA class II, D beta chain); Major histocompatibility complex class II beta chain BLB3, (similar to HLA class II, D beta chain); major histocompatibility complex class II DR-beta chain; major histocompatibility complex, class II, DQ alpha; major histocompatibility complex, class II, DQ alpha 1; major histocompatibility complex, class II, DQ alpha 1 precursor; major histocompatibility complex, class II, DQ alpha 2; major histocompatibility complex, class II, DQ alpha 5; major histocompatibility complex, class II, DQ alpha, type 1; major histocompatibility complex, class II, DQ alpha, type 3; major histocompatibility complex, class II, DQ beta; major histocompatibility complex, class II, DQ beta 1; major histocompatibility complex, class II, DR alpha; major histocompatibility complex, class II, DR alpha precursor; major histocompatibility complex, class II, DR beta 3; major histocompatibility complex, class II, DR beta 4 precursor; major histocompatibility complex, class II, DRB3; MHC class II antigen; MHC class II antigen alpha chain; MHC class II antigen beta chain; MHC class II antigen B-F minor heavy chain; MHC class II antigen BoLA-DRB3; MHC class II antigen precursor; MHC Class II beta 1 and 2 domains; MHC class II beta 1 domain; MHC class II beta chain; MHC class II beta chain 1; MHC class II beta chain 2; MHC class II B-L beta chain; MHC class II DLA DQ beta chain; MHC class II DLA DQA alpha chain; MHC class II DLA DQalpha chain; MHC class II DLA DRB1 beta chain; MHC class II DLA-DRB; MHC class II DLA-DR-beta-1; MHC class II DQA; MHC class II DQA precursor; MHC class II DQA1 precursor; MHC class II DQA2 (BoLA-DQA2); MHC class II DQA2 precursor; MHC class II DQ-alpha; MHC class II DQ-alpha chain; MHC class II DQB precursor; MHC class II DQ-beta chain; MHC class II DR alpha chain; MHC class II DR beta 1; MHC class II DR beta chain; MHC class II DR beta-chain; MHC class II DR-alpha; MHC class II DRB3; MHC class II DRB3 precursor; MHC class II DR-beta chain; MHC class II HLA-DQ-alpha; MHC class II leukocyte antigen BoLA-DRB3; MHC class IIA antigen; MHC DLA-DR-beta-1 precursor; MHC(BoLA) class II DR-beta chain; MHCII beta chain; S19; similar to MHC class II DQA2 (BoLA-DQA2); uncharacterized protein LOC101747454; Unknown (protein for MGC:127488); Unknown (protein for MGC:137100)
Gene Symbols DLA-DQA1, DLA-DQB1, HLA-DRB1
Host Species Rat
Immunogen Con A activated canine peripheral blood cells.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 474860, 474861, 474862
Target Species Canine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.