Learn More
Invitrogen™ MHC II Polymorphic Monoclonal Antibody (CI.13.I)

Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Guinea Pig MHC class II polymorphic antibody, clone CI-13.1 recognizes the MHC Class II epitope present on strain 13, but not strain 2 guinea pig cells. The antigen is also found in outbred Hartley guinea pigs.
Specifications
Specifications
| Antigen | MHC II Polymorphic |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | CI.13.I |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with 1% BSA and 0.1% sodium azide |
| Host Species | Mouse |
| Immunogen | Guinea pig peritoneal T-cells. |
| Quantity | 250 μL |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.