Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mi-2 beta Monoclonal Antibody (3F2/4), Active Motif™

Mouse Monoclonal Antibody
$132.55 - $397.65
Specifications
Antigen | Mi-2 beta |
---|---|
Clone | 3F2/4 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2788
![]() ![]() |
Active Motif
39696 |
10 μg |
Each for $132.55
|
|
|||||
50-199-2787
![]() ![]() |
Active Motif
39695 |
100 μg |
Each for $397.65
|
|
|||||
Description
The product of this gene belongs to the SNF2/RAD54 helicase family. It represents the main component of the nucleosome remodeling and deacetylase complex and plays an important role in epigenetic transcriptional repression. Patients with dermatomyositis develop antibodies against this protein.Specifications
Mi-2 beta | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
9530019N15Rik; AA617397; ATP-dependent helicase CHD4; BC005710; CHD4; CHD-4; chromodomain helicase DNA binding protein 4; Chromodomain-helicase-DNA-binding protein 4; D6Ertd380e; Mi-2; Mi-2 autoantigen; Mi-2 autoantigen 218 kDa protein; Mi-2 beta; MI2B; Mi-2b; Mi-2beta; Mi2-beta; mKIAA4075 | |
CHD4 | |
IgG | |
Protein G | |
Antibody |
3F2/4 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Q14839, Q6PDQ2 | |
107932, 1108 | |
This Mi-2 beta antibody was raised against a synthetic peptide derived from human Mi-2B. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
CHD4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title