Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIA Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299450PCP
Description
MIA Polyclonal antibody specifically detects MIA in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
MIA | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
CD-RAP, melanoma inhibitory activity, Melanoma inhibitory activity protein, melanoma-derived growth regulatory protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human MIA. | |
0.1 mL | |
Cancer | |
8190 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction