Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIA Polyclonal Antibody

Catalog No. PIPA535927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535927 Supplier Invitrogen™ Supplier No. PA535927
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A375, human MIA-transfected 293T. Predicted reactivity: Mouse (91%), Rat (93%), Rhesus Monkey (99%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Tumorigenesis is a process that is mediated by a network of membrane, cytosolic and nuclear associated factors, which regulate proliferation and cell-matrix interaction through signaling cascades. The phenotype of malignant melanomas in vivo depends on the global expression of stimulatory or inhibitory factors generated in both the tumors cells and their environment. One example, Melanoma inhibitory activity (cartilage-derived retinoic acid-sensitive protein (CD-RAP), MIA) is a Src homology 3 (SH3)-like domain containing protein that is secreted from chondrocytes and malignant melanoma cells. MIA is translated as a 131-amino acid pro-form and processed into a mature 107-amino acid protein after cleavage of a secretion signal. MIA is expressed during chondrogenesis and in mature chondrocytes, suggesting that MIA is necessary for normal cartilage cell phenotype. MIA mRNA is present in carcinomas of the colon, ovary, kidney, and head/neck, and may represent a marker to monitor melanomic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIA
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene MIA
Gene Accession No. Q16674
Gene Alias cartilage derived retinoic acid sensitive protein; cartilage-derived retinoic acid-sensitive protein; Cdrap; CD-RAP; melanoma inhibitory activity; melanoma inhibitory activity 1; melanoma inhibitory activity protein; melanoma inhibitory activity/condrocyte-derived retinoic acid sensitive protein homolog (MIA/CD-RAP); melanoma-derived growth regulatory protein; melanoma-inhibitory-activity; MIA; Mia1
Gene Symbols MIA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MIA. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8190
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.