Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIA3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521389

Catalog No. PIPA521389


Only null left
Add to Cart

Description

Description

Recommended positive controls: A549, HepG2. Predicted reactivity: Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Plays a role in the transport of cargos that are too large to fit into COPII-coated vesicles and require specific mechanisms to be incorporated into membrane-bound carriers and exported from the endoplasmic reticulum. This protein is required for collagen VII (COL7A1) secretion by loading COL7A1 into transport carriers. It may participate in cargo loading of COL7A1 at endoplasmic reticulum exit sites by binding to COPII coat subunits Sec23/24 and guiding SH3-bound COL7A1 into a growing carrier. Does not play a role in global protein secretion and is apparently specific to COL7A1 cargo loading. However, it may participate in secretion of other proteins in cells that do not secrete COL7A1. It is also specifically required for the secretion of lipoproteins by participating in their export from the endoplasmic reticulum (PubMed:27138255, PubMed:19269366). Required for correct assembly of COPII coat components at endoplasmic reticulum exit sites (ERES) and for the localization of SEC16A and membrane-bound ER-resident complexes consisting of MIA2 and PREB/SEC12 to ERES (PubMed:28442536). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MIA3
Polyclonal
Unconjugated
MIA3
9130229H14Rik; A930039G15Rik; ARNT; AU022327; B230399H06Rik; C219-reactive peptide; C80126; D320; Gm1525; Kiaa0268; melanoma inhibitory activity 3; melanoma inhibitory activity family member 3; melanoma inhibitory activity family, member 3; melanoma inhibitory activity protein 3; MIA family member 3, ER export factor; Mia3; RP11-378J18.4; Tango; TANGO1; transport and Golgi organization protein 1; Transport and Golgi organization protein 1 homolog; UNQ6077; UNQ6077/PRO20088
Rabbit
Antigen Affinity Chromatography
RUO
375056
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.3 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q5JRA6
MIA3
Recombinant fragment corresponding to a region within amino acids 1 and 150 of MIA3 (Uniprot ID#Q5JRA6).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.