Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563209
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563209 Supplier Invitrogen™ Supplier No. PA563209
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSKQGKPQSA TDYSDPDNVD DGLFIVDIPK TNNDKEVNAE HHIKGKGRGV QESKRGLVQD ETELEDENQE GMTVHSSVHS NNLNSMPA Highest antigen sequence identity to the following orthologs: Mouse - 25%, Rat - 26%.

Plays a role in the transport of cargos that are too large to fit into COPII-coated vesicles and require specific mechanisms to be incorporated into membrane-bound carriers and exported from the endoplasmic reticulum. This protein is required for collagen VII (COL7A1) secretion by loading COL7A1 into transport carriers. It may participate in cargo loading of COL7A1 at endoplasmic reticulum exit sites by binding to COPII coat subunits Sec23/24 and guiding SH3-bound COL7A1 into a growing carrier. Does not play a role in global protein secretion and is apparently specific to COL7A1 cargo loading. However, it may participate in secretion of other proteins in cells that do not secrete COL7A1. It is also specifically required for the secretion of lipoproteins by participating in their export from the endoplasmic reticulum (PubMed:27138255, PubMed:19269366). Required for correct assembly of COPII coat components at endoplasmic reticulum exit sites (ERES) and for the localization of SEC16A and membrane-bound ER-resident complexes consisting of MIA2 and PREB/SEC12 to ERES (PubMed:28442536). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIA3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MIA3
Gene Accession No. Q5JRA6
Gene Alias 9130229H14Rik; A930039G15Rik; ARNT; AU022327; B230399H06Rik; C219-reactive peptide; C80126; D320; Gm1525; Kiaa0268; melanoma inhibitory activity 3; melanoma inhibitory activity family member 3; melanoma inhibitory activity family, member 3; melanoma inhibitory activity protein 3; MIA family member 3, ER export factor; Mia3; RP11-378J18.4; Tango; TANGO1; transport and Golgi organization protein 1; Transport and Golgi organization protein 1 homolog; UNQ6077; UNQ6077/PRO20088
Gene Symbols MIA3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MIA3. Recombinant protein control fragment (Product #RP-99801).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375056
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.