Learn More
MIA3/TANGO Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304859AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and contains a predicted leucine zipper motif typical of many transcription factors.Specifications
MIA3/TANGO | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q5JRA6 | |
MIA3 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MIA3 | |
RP11-378J18.4, ARNT, D320, TANGO, TANGO1, UNQ6077, C219 reactive peptide, C219-reactive peptide, melanoma inhibitory activity protein 3, transport and Golgi organization protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
375056 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.