Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mianserin hydrochloride

Catalog No. aaj61570mc
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61570MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61570MC Supplier Thermo Scientific Chemicals Supplier No. J61570MC
Only null left
Add to Cart

CAS: 21535-47-7 | C18H21ClN2 | 300.83 g/mol

Mianserin hydrochloride is used as an antidepressant, antagonist, inverse agonist at 5-HT2 serotonin receptors, also blocks the H1 histamine receptor and the α2 adrenoceptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mianserin hydrochloride is used as an antidepressant, antagonist, inverse agonist at 5-HT2 serotonin receptors, also blocks the H1 histamine receptor and the α2 adrenoceptor.

Solubility
Soluble in water, ethano
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21535-47-7
Molecular Formula C18H21ClN2
Molecular Weight (g/mol) 300.83
MDL Number MFCD00055072
InChI Key YNPFMWCWRVTGKJ-UHFFFAOYNA-N
Synonym mianserin hydrochloride, mianserine hydrochloride, mianserin hcl, athymil, bolvidon, tolvon, tolvin, org gb 94, lerivon
PubChem CID 68551
ChEBI CHEBI:31843
IUPAC Name hydrogen 5-methyl-2,5-diazatetracyclo[13.4.0.0²,⁷.0⁸,¹³]nonadeca-1(19),8,10,12,15,17-hexaene chloride
SMILES [H+].[Cl-].CN1CCN2C(C1)C1=CC=CC=C1CC1=CC=CC=C21

Specifications

Color White
Quantity 100 mg
Merck Index 14,6172
Solubility Information Soluble in water,ethanol,methanol,and dimethylformamide.
Formula Weight 300.83
Physical Form Powder
Chemical Name or Material Mianserin hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 244-426-7
RTECSNumber HP8780000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.