Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mianserin Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M26231G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26231G 1 g
M2623200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26231G Supplier TCI America Supplier No. M26231G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21535-47-7
Molecular Formula C18H21ClN2
Molecular Weight (g/mol) 300.83
MDL Number MFCD00055072
InChI Key YNPFMWCWRVTGKJ-UHFFFAOYNA-N
Synonym mianserin hydrochloride, mianserine hydrochloride, mianserin hcl, athymil, bolvidon, tolvon, tolvin, org gb 94, lerivon
PubChem CID 68551
ChEBI CHEBI:31843
IUPAC Name hydrogen 5-methyl-2,5-diazatetracyclo[13.4.0.0²,⁷.0⁸,¹³]nonadeca-1(19),8,10,12,15,17-hexaene chloride
SMILES [H+].[Cl-].CN1CCN2C(C1)C1=CC=CC=C1CC1=CC=CC=C21

Specifications

Melting Point 284°C
Color White-Yellow
Quantity 1 g
Formula Weight 300.83
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Mianserin Hydrochloride
RTECSNumber HP8780000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.