Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mib1/Mindbomb Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982201
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2201 Supplier Bioss Supplier No. BS12391R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MIB is an E3 ubiquitin-protein ligase that mediates ubiquitination of Delta receptors, which act as ligands of Notch proteins. This protein positively regulates the Delta-mediated Notch signaling by ubiquitinating the intracellular domain of Delta, leading to endocytosis of Delta receptors. MIB probably mediates ubiquitination and subsequent proteasomal degradation of DAPK1, thereby antagonizing anti-apoptotic effects of DAPK1 to promote TNF-induced apoptosis.

Specifications

Antigen Mib1/Mindbomb
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MIB1
Gene Accession No. Q86YT6
Gene Alias DAPK-interacting protein 1; DAPK-interacting protein-1; Dip1; DIP-1; E3 ubiquitin-protein ligase MIB1; E430019M12Rik; Kiaa1323; LVNC7; Mib; MIB1; Mind bomb homolog 1; mind bomb-1; mindbomb E3 ubiquitin protein ligase 1; mindbomb homolog 1; mindbomb homolog 1 (Drosophila); mKIAA1323; RING-type E3 ubiquitin transferase MIB1; skeletrophin; ubiquitin ligase mind bomb; Zinc finger ZZ type with ankyrin repeat domain protein 2; ZZANK2; ZZZ6
Gene Symbols MIB1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Mib1/Mindbomb.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57534
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.