Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIB1 Mouse anti-Human, Monkey, Clone:6A9C9, Abnova™

Catalog No. 89334490 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-490 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-490 Supplier Abnova Corporation Supplier No. MAB17826
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human MIB1.

Specifications

Antigen MIB1
Applications ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 6A9C9
Conjugate Unconjugated
Description mindbomb homolog 1 (Drosophila)
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunocytochemistry (1:200-1:1000)
Immunohistochemistry
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene MIB1
Gene Alias DIP-1, DKFZp686I0769, DKFZp761M1710, FLJ90676, MGC129659, MGC129660, MIB, ZZANK2, ZZZ6
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 6-221 of human MIB1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 57534
Target Species Human, Monkey
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.