Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597126 Supplier Invitrogen™ Supplier No. PA597126
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSEGDTPLHD AISKKRDDIL AVLLEAGADV TITNNNGFNA LHHAALRGNP SAMRVLLSKL PRPWIVDEKK DDGYTALHLA ALNNHVEVAE LLVHQGNANL DIQNVNQQTA LHLAVERQHT QIVRLLVRAG AKLDIQDKDG DTPLHEALRH HTLSQLRQLQ DMQDVGKVDA AWEPSKNTLI MGLGTQGAEK KSAASIACFL AANGADLSIR NKKGQSPLDL CPDPNLCKAL AKCHKEKVSG QVGSRSPSMI SNDSETLEEC MVCSDMKRDT LFGPCGHIAT CSLCSPRVKK CLICKEQVQS R; Positive Samples: U-87MG, A-549, HepG2, Mouse stomach, Mouse lung, Mouse kidney, Rat testis; Cellular Location: Cell membrane, Cytoplasm, centriolar satellite, centrosome, cytoskeleton, microtubule organizing center.

MIB is an E3 ubiquitin-protein ligase that mediates ubiquitination of Delta receptors, which act as ligands of Notch proteins. This protein positively regulates the Delta-mediated Notch signaling by ubiquitinating the intracellular domain of Delta, leading to endocytosis of Delta receptors. MIB probably mediates ubiquitination and subsequent proteasomal degradation of DAPK1, thereby antagonizing anti-apoptotic effects of DAPK1 to promote TNF-induced apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIB1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene MIB1
Gene Accession No. Q80SY4, Q86YT6
Gene Alias DAPK-interacting protein 1; DAPK-interacting protein-1; DIP1; DIP-1; E3 ubiquitin-protein ligase MIB1; E430019M12Rik; KIAA1323; LVNC7; MIB; MIB1; Mind bomb homolog 1; mind bomb-1; mindbomb E3 ubiquitin protein ligase 1; mindbomb homolog 1; mindbomb homolog 1 (Drosophila); mKIAA1323; RING-type E3 ubiquitin transferase MIB1; skeletrophin; ubiquitin ligase mind bomb; zinc finger ZZ type with ankyrin repeat domain protein 2; ZZANK2; ZZZ6
Gene Symbols MIB1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 560-860 of human MIB1/DIP1 (NP_0658251).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225164, 307594, 57534
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.