Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MIB1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142904
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
MIB is an E3 ubiquitin-protein ligase that mediates ubiquitination of Delta receptors, which act as ligands of Notch proteins. This protein positively regulates the Delta-mediated Notch signaling by ubiquitinating the intracellular domain of Delta, leading to endocytosis of Delta receptors. MIB probably mediates ubiquitination and subsequent proteasomal degradation of DAPK1, thereby antagonizing anti-apoptotic effects of DAPK1 to promote TNF-induced apoptosis.
Specifications
MIB1 | |
Polyclonal | |
Unconjugated | |
MIB1 | |
DAPK-interacting protein 1; DAPK-interacting protein-1; DIP1; DIP-1; E3 ubiquitin-protein ligase MIB1; E430019M12Rik; KIAA1323; LVNC7; MIB; MIB1; Mind bomb homolog 1; mind bomb-1; mindbomb E3 ubiquitin protein ligase 1; mindbomb homolog 1; mindbomb homolog 1 (Drosophila); mKIAA1323; RING-type E3 ubiquitin transferase MIB1; skeletrophin; ubiquitin ligase mind bomb; zinc finger ZZ type with ankyrin repeat domain protein 2; ZZANK2; ZZZ6 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
57534 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q86YT6 | |
MIB1 | |
Peptide with sequence C-DKDNTNVNADVQKLQ. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction