Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIB1 Recombinant Rabbit Monoclonal Antibody (JG69-34)
GREENER_CHOICE

Catalog No. PIMA534758
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534758 Supplier Invitrogen™ Supplier No. MA534758
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: HUVEC, SH-SY-5Y, SiHa, human appendix tissue, human kidney tissue.

MIB is an E3 ubiquitin-protein ligase that mediates ubiquitination of Delta receptors, which act as ligands of Notch proteins. This protein positively regulates the Delta-mediated Notch signaling by ubiquitinating the intracellular domain of Delta, leading to endocytosis of Delta receptors. MIB probably mediates ubiquitination and subsequent proteasomal degradation of DAPK1, thereby antagonizing anti-apoptotic effects of DAPK1 to promote TNF-induced apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIB1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JG69-34
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MIB1
Gene Accession No. Q86YT6
Gene Alias DAPK-interacting protein 1; DAPK-interacting protein-1; DIP1; DIP-1; E3 ubiquitin-protein ligase MIB1; E430019M12Rik; KIAA1323; LVNC7; MIB; MIB1; Mind bomb homolog 1; mind bomb-1; mindbomb E3 ubiquitin protein ligase 1; mindbomb homolog 1; mindbomb homolog 1 (Drosophila); mKIAA1323; RING-type E3 ubiquitin transferase MIB1; skeletrophin; ubiquitin ligase mind bomb; zinc finger ZZ type with ankyrin repeat domain protein 2; ZZANK2; ZZZ6
Gene Symbols MIB1
Host Species Rabbit
Immunogen Recombinant protein within Human Mib1 aa 2-93.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57534
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.