Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MIB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
$249.00 - $605.00
Specifications
| Antigen | MIB2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119285
![]() |
Invitrogen™
PA5119285 |
200 μL |
Each for $605.00
|
|
|||||
|
PIPA5121774
|
Invitrogen™
PA5121774 |
20 μL |
Each for $249.00
|
|
|||||
Description
MIB2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
E3 ubiquitin-protein ligase that mediates ubiquitination of Delta receptors, which act as ligands of Notch proteins. Positively regulates the Delta-mediated Notch signaling by ubiquitinating the intracellular domain of Delta, leading to endocytosis of Delta receptors.Specifications
| MIB2 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| MIB2 | |
| 2210008I11Rik; Dyslike; dystrophin-like protein; E3 ubiquitin-protein ligase MIB2; EC 6.3.2.13; FLJ20648; FLJ41406; Mib2; Mind bomb homolog 2; mind bomb-2; mindbomb E3 ubiquitin protein ligase 2; mindbomb homolog 2; mindbomb homolog 2 (Drosophila); novel zinc finger protein; Novelzin; putative NF-kappa-B-activating protein 002N; RBSC-skeletrophin; RBSC-skeletrophin/dystrophin-like polypeptide; RING-type E3 ubiquitin transferase MIB2; skd; Skeletrophin; zinc finger ZZ type with ankyrin repeat domain protein 1; zinc finger, ZZ type with ankyrin repeat domain 1; Zzank1; ZZZ5 | |
| MIB2 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8R516 | |
| 76580 | |
| Recombinant protein Mindbomb Homolog 2. The antigen corresponds to amino acid range 546-784 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title