Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100871
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100871 Supplier Invitrogen™ Supplier No. PA5100871
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MIB2.

E3 ubiquitin-protein ligase that mediates ubiquitination of Delta receptors, which act as ligands of Notch proteins. Positively regulates the Delta-mediated Notch signaling by ubiquitinating the intracellular domain of Delta, leading to endocytosis of Delta receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIB2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MIB2
Gene Accession No. Q68LP1, Q8R516, Q96AX9
Gene Alias 2210008I11Rik; Dyslike; dystrophin-like protein; E3 ubiquitin-protein ligase MIB2; EC 6.3.2.13; FLJ20648; FLJ41406; Mib2; Mind bomb homolog 2; mind bomb-2; mindbomb E3 ubiquitin protein ligase 2; mindbomb homolog 2; mindbomb homolog 2 (Drosophila); novel zinc finger protein; Novelzin; putative NF-kappa-B-activating protein 002N; RBSC-skeletrophin; RBSC-skeletrophin/dystrophin-like polypeptide; RING-type E3 ubiquitin transferase MIB2; skd; Skeletrophin; zinc finger ZZ type with ankyrin repeat domain protein 1; zinc finger, ZZ type with ankyrin repeat domain 1; Zzank1; ZZZ5
Gene Symbols MIB2
Host Species Rabbit
Immunogen A synthesized peptide derived from human MIB2(Accession Q96AX9), corresponding to amino acid residues F693-P743.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142678, 474147, 76580
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.