Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Mibefradil dihydrochloride
GREENER_CHOICE

Catalog No. 219810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-981-0 10 mg
21-985-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-981-0 Supplier Tocris Bioscience™ Supplier No. 2198/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

CaV3.x blocker

Mibefradil dihydrochloride is a Ca2+ channel blocker with moderate selectivity for T-type Ca2+ channels displaying IC50 values of 2.7 μM and 18.6 μM for T-type and L-type channels respectively. Antihypertensive agent.

Specifications

Quantity 10 mg
Product Type Mibefradil dihydrochloride
Purity >97%
Molecular Weight (g/mol) 568.56
Inhibitors Mibefradil dihydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.