Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIC-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560428 Supplier Invitrogen™ Supplier No. PA560428
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CLLLSLESFY PPAHQLSLDM LKRLSTANDE IVEVLLSKHQ VLAALRFIRG IGGHDNISAR KFLDAAKQTE DNMLFYTIFR FFEQRNQRLR GSPNFTPGE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

Colon cancer-associated protein Mic1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIC-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rmc1
Gene Accession No. Q96DM3
Gene Alias 2400010D15Rik; 3110002H16Rik; C18orf8; chromosome 18 open reading frame 8; colon cancer-associated protein Mic1; colon cancer-associated protein Mic1 homolog; HsT2591; macrophage inhibitory cytokine 1; Mic1; Mic-1; Regulator of MON1-CCZ1 complex; RGD1311805; RIKEN cDNA 3110002H16 gene; RMC1; similar to RIKEN cDNA 2400010D15; uncharacterized protein C18orf8; uncharacterized protein C18orf8 homolog; WD repeat-containing protein 98; WDR98
Gene Symbols Rmc1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MIC-1. Recombinant protein control fragment (Product #RP-98280).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29919
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.