Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIC A/B Mouse anti-Human, Brilliant Violet 421, Clone: 6D4, BD Optibuild™

Catalog No. BDB742322 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742322 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742322 Supplier BD Biosciences Supplier No. 742322
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 6D4 monoclonal antibody specifically binds to the human MHC class I polypeptide-related sequence A (MICA, aka /PERB11.1) and B (MICB. aka /PERB11.2) proteins. These ñ70 kDa transmembrane glycoproteins are homologs of the major histocompatibility complex class I molecules although they lackassociation with β2 microglobulin. The MHC class I-related MICA and MICB chains are expressed by some gut epithelial cells in vivo. MICA and MICB expression by other epithelial cells and cell types, including fibroblasts and endothelial cells, is induced by stress, eg, stress caused by bacterial and viral infections, autoimmunity or cellular transformation. Epithelial cell expression of MICA and MICB has also been detected in transplanted kidneys and pancreas that show histological signs of rejection and or cellular injury. This suggests their potential role in transplant immunopathology. MICA and MICB are ligands for NKG2D (CD314), an activating receptor expressed by natural killer (NK) cells,γδ T cells,CD8+ andsome CD4+ αβ T cells. The 6D4 antibody reportedly blocks NKG2D-positive NK cell- and T cell-mediated cytotoxicity against MICA/B-positive target cells.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIC A/B
Classification Monoclonal
Clone 6D4
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias MHC class I polypeptide-related sequence A/B, PERB11.1/ PERB11.2
Host Species Mouse
Immunogen MICA transfected cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.