Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mic1 Antibody (OTI4E4), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272668FR
Description
Mic1 Monoclonal specifically detects Mic1 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
Mic1 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
RMC1 | |
Full length recombinant protein of human Mic1 (NP_037458) produced in HEK293T cells. | |
0.1 mL | |
29919.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
OTI4E4 | |
Western Blot, Flow Cytometry | |
chromosome 18 open reading frame 8, Colon cancer-associated protein Mic1, FLJ23453, HsT2591, Mic-1, MIC1colon cancer associated protein Mic1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction