Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mic1 Antibody (OTI4E4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP201949
Description
Mic1 Monoclonal specifically detects Mic1 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
Mic1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Flow Cytometry 1:100 | |
RMC1 | |
Full length human recombinant protein of human C18orf8(NP_037458) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
29919 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry | |
OTI4E4 | |
Unconjugated | |
chromosome 18 open reading frame 8, Colon cancer-associated protein Mic1, FLJ23453, HsT2591, Mic-1, MIC1colon cancer associated protein Mic1 | |
Mouse | |
74.8 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction