Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MICA Antibody (159227), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1300PECY55
Description
MICA Monoclonal antibody specifically detects MICA in Human samples. It is validated for Flow CytometrySpecifications
MICA | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
159227 | |
Flow Cytometry | |
FLJ60820, MGC111087, PERB11.1 | |
BaF3 mouse pro-B cell line transfected with human MICA | |
0.1 mL | |
Biologically Active Proteins | |
100507436 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction