Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MICA/B Antibody (6D4), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP276811MFV610
Description
MICA/B Monoclonal antibody specifically detects MICA/B in Human samples. It is validated for Flow CytometrySpecifications
MICA/B | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
100507436 | |
Store at 4C in the dark. | |
IgG2 α |
Flow Cytometry | |
6D4 | |
Flow Cytometry | |
MHC class I chain-related protein A, MHC class I chain-related protein B, MIC, MICA, MICB, PERB11, PERB11.1, PERB11.2 | |
Transfected C1R cells expressing MICA | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction