Learn More
Invitrogen™ MICA/B Monoclonal Antibody (6D4), APC

Description
This antibody recognizes a common extracellular epitope on MICA and MICB glycoproteins, transmembrane ligands of CD314 (NKG2D), and is able to block CD314-mediated activation of NK cells and cytotoxic T cells.
Specifications
Specifications
| Antigen | MICA/B |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 6D4 |
| Concentration | 100 μg/mL |
| Conjugate | APC |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| Gene | MICA |
| Gene Accession No. | Q29980, Q29983 |
| Gene Alias | HLA class I antigen; MHC class I chain-related protein A; MHC class I chain-related protein B; MHC class I mic-B antigen; MHC class I polypeptide-related sequence A; MHC class I polypeptide-related sequence B; MHC class I related sequence A; MHC class I-like molecule PERB11.2-IMX; MICA; MIC-A; MICB; MIC-B; PERB11.1; PERB11.2; sMICA; sMICB; soluble MICA; soluble MICB; stress inducible class I homolog; truncated MHC class I polypeptide-related sequence A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.