Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MICA Monoclonal Antibody (OTI2C3), TrueMAB™, OriGene
SDP

Catalog No. 502543672
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3672 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3672 Supplier OriGene Supplier No. CF813273
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MICA Monoclonal antibody specifically detects MICA in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen MICA
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2C3
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CD47
Gene Accession No. Q08722
Gene Alias DAMA-345G11.2; HLA class I antigen; MHC class I chain-related protein A; MHC class I polypeptide-related sequence A; MHC class I related sequence A; MICA; MIC-A; PERB11.1; stress inducible class I homolog; truncated MHC class I polypeptide-related sequence A
Host Species Mouse
Immunogen Full length human recombinant protein of human CD47 (NP_001768) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 961
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.