Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MICA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579671
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Raji whole cell, human SGC-7901 whole cell, human SW620 whole cell, huamn Jurkat whole cell.
Major histocompatibility complex (MHC) class I proteins are ubiquitously expressed and mediate the recognition of intracellular antigens by cytotoxic T cells. A related family, termed the MHC class I chain-related (MIC) proteins are recognized by NKG2D, a receptor on NK and T cells, and promote anti-tumor activity. MICA, a member of the MIC family, is widely expressed on many tumors, and it is the MICA/NKG2D interaction that is thought to stimulate the anti-tumor reactivity by T lymphocytes. Both MICA and MICB mRNA are widely expressed in normal tissues, with MICA being present in virtually every tissue except the nervous system, suggesting that MIC protein expression may only be one component of the anti-tumor activity of the immune system.
Specifications
MICA | |
Polyclonal | |
Unconjugated | |
MICA | |
DAMA-345G11.2; HLA class I antigen; MHC class I chain-related protein A; MHC class I polypeptide-related sequence A; MHC class I related sequence A; MICA; MIC-A; PERB11.1; stress inducible class I homolog; truncated MHC class I polypeptide-related sequence A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100507436 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 4 mg trehalose and 0.01 mg sodium azide | |
Q29983 | |
MICA | |
A synthetic peptide corresponding to a sequence at the C-terminus of human MICA (304-334aa QSHWQTFHVSAVAAAAKFVEIIFYVRCCKKK). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction