Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MICA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589517
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589517 Supplier Invitrogen™ Supplier No. PA589517
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPHSLRYNLT VLSWDGSVQS GFLTEVHLDG QPFLRCDRQK CRAKPQGQWA EDVLGNKTWD RETRDLTGNG KDLRMTLAHI KDQKEGLHSL QEIRVCEIHE DNSTRSSQHF YYDGELFLSQ NLETKEWTMP QSSRAQTLAM NVRNFLKEDA MKTKTHYHAM HADCLQELRR YLKSGVVLRR TVPPMVNVTR SEASEGNITV TCRASGFYPW NITLSWRQDG VSLSHDTQQW GDVLPDGNGT YQTWVATRIC QGEEQRFTCY MEHSGNHSTH PVPSGKVLVL QSHW; Positive Samples: MCF7, SW480, NCI-H460, HepG2, HeLa, Mouse kidney, Mouse ovary, Rat kidney; Cellular Location: Cell membrane, Cytoplasm, Single-pass type I membrane protein.

Major histocompatibility complex (MHC) class I proteins are ubiquitously expressed and mediate the recognition of intracellular antigens by cytotoxic T cells. A related family, termed the MHC class I chain-related (MIC) proteins are recognized by NKG2D, a receptor on NK and T cells, and promote anti-tumor activity. MICA, a member of the MIC family, is widely expressed on many tumors, and it is the MICA/NKG2D interaction that is thought to stimulate the anti-tumor reactivity by T lymphocytes. Both MICA and MICB mRNA are widely expressed in normal tissues, with MICA being present in virtually every tissue except the nervous system, suggesting that MIC protein expression may only be one component of the anti-tumor activity of the immune system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MICA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MICA
Gene Accession No. Q29983
Gene Alias DAMA-345G11.2; HLA class I antigen; MHC class I chain-related protein A; MHC class I polypeptide-related sequence A; MHC class I related sequence A; MICA; MIC-A; PERB11.1; stress inducible class I homolog; truncated MHC class I polypeptide-related sequence A
Gene Symbols MICA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-307 of human MICA (NP_0002381).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100507436
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.