Learn More
Eisco™ Mica Schist Specimen
SCI_ED
Catalog No. S24795 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24795 12/Pk. 2 to 3 cm
S24794 1 Ea. 2 to 3 cm
S26680 10/Pk. 2 to 3 cm
S26654 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24795 Supplier Eisco™ Supplier No. ESNG0062PK12
Add to Cart
Add to Cart
Mica Schist Specimen for use in lessons on metamorphic rock properties, classification, and identification.
  • Distinctly altered mineral crystals are ideal for students studying metasomatism in metamorphic rocks
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Product of moderate grade metamorphism from granite
  • Has undergone moderate levels of alteration
  • Schist is good for demonstrating effect of metamorphism on various minerals
Specifications
Product Type Metamorphic Rock
For Use With (Application) Lessons on metamorphic rock properties, classification, and identification
Packaging Clear labeled and barcoded bag
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Specimen Type Mica Schist