Learn More
Eisco™ Mica Schist Specimen
SCI_ED
Catalog No. S26680 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S26680 10/Pk. 2 to 3 cm
S24794 1 Ea. 2 to 3 cm
S24795 12/Pk. 2 to 3 cm
S26654 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S26680 Supplier Eisco™ Supplier No. ESNG0062PK10
Add to Cart
Add to Cart
Mica Schist Specimen for use in lessons on metamorphic rock properties, classification, and identification.
  • Distinctly altered mineral crystals are ideal for students studying metasomatism in metamorphic rocks
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Product of moderate grade metamorphism from granite
  • Has undergone moderate levels of alteration
  • Schist is good for demonstrating effect of metamorphism on various minerals
Specifications
Product Type Metamorphic Rock
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Specimen Type Mica Schist