Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MICAL1 Antibody (OTI6H4), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272670PECY7
Description
MICAL1 Monoclonal specifically detects MICAL1 in Human samples. It is validated for Flow Cytometry.Specifications
MICAL1 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
MICAL1 | |
Full length recombinant protein of human MICAL1 (NP_073602) produced in HEK293T cells. | |
0.1 mL | |
64780.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI6H4 | |
Flow Cytometry | |
CasL interacting molecule, DKFZp434B1517, FLJ11937, FLJ21739, MICALMolecule interacting with CasL protein 1, microtubule associated monoxygenase, calponin and LIM domain containing 1, NEDD9 interacting protein with calponin homology and LIM domains, NEDD9-interacting protein with calponin homology and LIM domains, NICALMICAL-1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction