Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MICAL1 Monoclonal Antibody (OTI5C3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501843

Description
May be a cytoskeletal regulator that connects NEDD9 to intermediate filaments.Specifications
MICAL1 | |
Monoclonal | |
0.76 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8TDZ2 | |
MICAL1 | |
Full length human recombit protein of human MICAL1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI5C3 | |
Unconjugated | |
MICAL1 | |
MICAL, MICAL-1, NICAL | |
Mouse | |
Affinity Chromatography | |
RUO | |
64780 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction