Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MICAL1 Monoclonal Antibody (OTI6H10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501845S


Description
MICAL1 Monoclonal antibody specifically detects MICAL1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
MICAL1 | |
Monoclonal | |
0.71 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8TDZ2 | |
Mouse | |
Affinity chromatography | |
RUO | |
64780 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI6H10 | |
Unconjugated | |
MICAL1 | |
[F-actin]-monooxygenase MICAL1; CasL interacting molecule MICAL; MICAL; MICAL1; MICAL-1; microtubule associated monooxygenase, calponin and LIM domain containing 1; microtubule associated monoxygenase, calponin and LIM domain containing 1; mMical1; molecule interacting with CasL protein 1; NEDD9 interacting protein with calponin homology and LIM domains; NEDD9-interacting protein with calponin homology and LIM domains; NICAL; protein-methionine sulfoxide oxidase MICAL1 | |
Full length human recombinant protein of human MICAL1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction