Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MICAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556388
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556388 Supplier Invitrogen™ Supplier No. PA556388
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRDLYDVLAK EPVQRNNDKT DTGMPATGSA GTQEELLRWC QEQTAGYPGV HVSDLSSSWA DGLALCALVY RLQPGLLEPS ELQGLGALEA TAWALKV Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 72%.

May be a cytoskeletal regulator that connects NEDD9 to intermediate filaments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MICAL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MICAL1
Gene Accession No. Q8TDZ2
Gene Alias [F-actin]-monooxygenase MICAL1; CasL interacting molecule MICAL; MICAL; MICAL1; MICAL-1; microtubule associated monooxygenase, calponin and LIM domain containing 1; microtubule associated monoxygenase, calponin and LIM domain containing 1; mMical1; molecule interacting with CasL protein 1; NEDD9 interacting protein with calponin homology and LIM domains; NEDD9-interacting protein with calponin homology and LIM domains; NICAL; protein-methionine sulfoxide oxidase MICAL1
Gene Symbols MICAL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MICAL1. Recombinant protein control fragment (Product #RP-94852).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64780
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.