Learn More
Invitrogen™ MICAL1 Polyclonal Antibody

Description
Positive Samples: A-549, HT-29, Raji; Cellular Location: Cytoplasm, cytoskeleton.
Specifications
Specifications
| Antigen | MICAL1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.75 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | MICAL1 |
| Gene Accession No. | Q8TDZ2, Q8VDP3 |
| Gene Alias | [F-actin]-monooxygenase MICAL1; CasL interacting molecule MICAL; MICAL; MICAL1; MICAL-1; microtubule associated monooxygenase, calponin and LIM domain containing 1; microtubule associated monoxygenase, calponin and LIM domain containing 1; mMical1; molecule interacting with CasL protein 1; NEDD9 interacting protein with calponin homology and LIM domains; NEDD9-interacting protein with calponin homology and LIM domains; NICAL; protein-methionine sulfoxide oxidase MICAL1 |
| Gene Symbols | MICAL1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.