Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MICAL1 Polyclonal Antibody
MICAL1 Polyclonal Antibody
Supplier: Thermo Scientific PA598764
Description
MICAL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Specifications
MICAL1 | |
Polyclonal | |
Unconjugated | |
MICAL1 | |
[F-actin]-monooxygenase MICAL1; CasL interacting molecule MICAL; MICAL; MICAL1; MICAL-1; microtubule associated monooxygenase, calponin and LIM domain containing 1; microtubule associated monoxygenase, calponin and LIM domain containing 1; mMical1; molecule interacting with CasL protein 1; NEDD9 interacting protein with calponin homology and LIM domains; NEDD9-interacting protein with calponin homology and LIM domains; NICAL; protein-methionine sulfoxide oxidase MICAL1 | |
Rabbit | |
Protein G | |
RUO | |
64780 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.76 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8TDZ2 | |
MICAL1 | |
Recombinant Human [F-actin]-methionine sulfoxide oxidase MICAL1 protein (806-1067AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction