Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MICAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556474
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556474 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556474 Supplier Invitrogen™ Supplier No. PA556474
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVTTGKEMAS AQEPDKLSMV MYLSKFYELF RGTPLRPVDS WRKNYGENAD LSLAKSSISN NYLNLTFPRK RTPRVDGQTG ENDMNKRRRK GFTNLDEPSN FSSRSLGSNQ ECGSSKEGGN QNKVKSM Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.

MICALs (Molecules Interacting with CasL) are atypical multidomain flavoenzymes with diverse cellular functions. There are three known isoforms, MICAL1, MICAL2 and MICAL3, as well as the MICAL-like proteins MICAL-L1 and MICAL-L2. MICAL2 has three conserved domains: an N-terminal flavin adenine dinucleotide (FAD) binding domain, a calponin homology (CH) domain and a Lin11, Isl-1 and Mec-3 (LIM) domain. It has been demonstrated that MICAL2 could regulate actin stress fibers and is required for normal actin organization. In addition, MICAL2-PV, a novel splicing variants of MICAL2, has been reported to be involved in cancer progression of prostate cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MICAL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mical2
Gene Accession No. O94851
Gene Alias [F-actin]-monooxygenase MICAL2; 5330438E18Rik; 9530064J02; flavoprotein oxidoreductase MICAL2; Kiaa0750; Mical2; MICAL-2; MICAL2PV1; MICAL2PV2; microtubule associated monooxygenase, calponin and LIM domain containing 2; microtubule associated monoxygenase, calponin and LIM domain containing 2; mKIAA0750; mMical2; molecule interacting with CasL protein 2; protein MICAL-2; protein-methionine sulfoxide oxidase MICAL2; RGD1311773
Gene Symbols Mical2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MICAL2. Recombinant protein control fragment (Product #RP-88764).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9645
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.