Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MICAL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551431
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551431 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551431 Supplier Invitrogen™ Supplier No. PA551431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVQSQSDTKD RLGSPLAVDE ALRRSDLVEE FWMKSAEIRR SLGLTPVDRS KGPEPSFPTP AFRPVSLKSY SVEKSPQDEG LHLLKPLSIP KRLGLPKPEG EPLSLPTPRS PSDRELRSAQ EERRELSSSS GLGLHGSSSN MKTL Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.

Monooxygenase that promotes depolymerization of F-actin by mediating oxidation of specific methionine residues on actin to form methionine-sulfoxide, resulting in actin filament disassembly and preventing repolymerization. In the absence of actin, it also functions as a NADPH oxidase producing H(2)O(2). Seems to act as Rab effector protein and plays a role in vesicle trafficking. Involved in exocytic vesicles tethering and fusion: the monooxygenase activity is required for this process and implicates RAB8A associated with exocytotic vesicles. Required for cytokinesis. Contributes to stabilization and/or maturation of the intercellular bridge independently of its monooxygenase activity. Promotes recruitment of Rab8 and ERC1 to the intercellular bridge, and together these proteins are proposed to function in timely abscission. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MICAL3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MICAL3
Gene Accession No. Q7RTP6
Gene Alias [F-actin]-monooxygenase MICAL3; flavoprotein oxidoreductase MICAL3; KIAA0819; KIAA1364; MICAL3; MICAL-3; microtubule associated monooxygenase, calponin and LIM domain containing 3; molecule interacting with CasL protein 3; protein MICAL-3; protein-methionine sulfoxide oxidase MICAL3
Gene Symbols MICAL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MICAL3. Recombinant protein control fragment (Product #RP-91479).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57553
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.