Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MICALL1 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP270017F
Description
MICALL1 Polyclonal specifically detects MICALL1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MICALL1 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
DKFZp686M2226, KIAA1668, MICAL-L1, MICAL-like 1, MICAL-like protein 1 | |
Rabbit | |
Affinity Purified | |
Signal Transduction | |
85377.0 | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
FITC | |
PBS with 0.05% Sodium Azide | |
MICALL1 | |
Synthetic peptide made to an internal portion of the human MICALL1 protein (between amino acids 200-280) [UniProt Q8N3F8] | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction