Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MICB Antibody (B-D54), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318102MFV500
Description
MICB Monoclonal antibody specifically detects MICB in Human samples. It is validated for Flow CytometrySpecifications
MICB | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
B-D54 | |
Flow Cytometry | |
MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog | |
Recombinant human MICB. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
0.1 mL | |
Biologically Active Proteins | |
4277 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction