Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MICB Antibody (MM0473-3C37), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211798APC
Description
MICB Monoclonal specifically detects MICB in Human samples. It is validated for Flow Cytometry.Specifications
MICB | |
Monoclonal | |
APC | |
NP_001170990 | |
MICB | |
Human recombinant MICB | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
MM0473-3C37 | |
Flow Cytometry | |
MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog | |
Mouse | |
Protein G purified | |
RUO | |
4277 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction