Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MICB Mouse anti-Human, Clone: B-D55, Novus Biologicals™

Mouse Monoclonal Antibody
$366.50 - $699.00
Specifications
Antigen | MICB |
---|---|
Clone | B-D55 |
Concentration | 1 mg/ml |
Dilution | ELISA |
Applications | ELISA |
Description
MICB Monoclonal antibody specifically detects MICB in Human samples. It is validated for ELISASpecifications
MICB | |
1 mg/ml | |
ELISA | |
Unconjugated | |
Mouse | |
Biologically Active Proteins | |
PBS. Sterile-filtered through 0.22 um. | |
4277 | |
IgG1 κ | |
Affinity purified |
B-D55 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog | |
Recombinant human MICB. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
Primary | |
Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title