Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MICB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579672
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579672 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579672 Supplier Invitrogen™ Supplier No. PA579672
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Jurkat whole cell, human K562 whole cell, human HEL whole cell, human A431 whole cell, human 293T whole cell, human SiHa whole cell, human HEPA1/6 whole cell. Flow: K562 cell.

MICB encodes the highly polymorphic MHC (HLA) class I chain-related gene B. The protein product is expressed on the cell surface. It is thought that MICB functions as a stress-induced antigen that is broadly recognized by intestinal epithelial gamma delta T cells. MICB is broadly recognized by NK cells and T cells with NKG2D receptor on their surface. The complex NKG2D-MICB results in MICB expressing cytolytic T cells and NK cells against epithelial tumor cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MICB
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene MICB
Gene Accession No. Q29980
Gene Alias MHC class I chain-related protein B; MHC class I mic-B antigen; MHC class I polypeptide-related sequence B; MHC class I-like located near the LRC, 2; MHC class I-like molecule PERB11.2-IMX; MHC I like leukocyte 2; MICB; MIC-B; Mill2; Mill2 gene for MHC class I-like located near the LRC, 2, exon 3, partial cds, strain:LEW.1lm1; PERB11.2; sMICB; soluble MICB; stress inducible class I homolog
Gene Symbols MICB
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human MICB (23-48aa AEPHSLRYNLMVLSQDESVQSGFLAE).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4277
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.