Learn More
MilliporeSigma™ Supelco™ Michel-Miller Column Stopper Ace-Thred
Catalog No. 111010583 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Thread Size:
Ace-Thred No. 15
Ace-Thred No. 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size
11-101-0583 Ace-Thred No. 25
11-101-0885 Ace-Thred No. 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-0583 Supplier MilliporeSigma™ Supelco™ Supplier No. Z1798251EA
Add to Cart
Add to Cart
PTFE for use at either end of any chromatography column with Ace-Threds. Makes a leak-tight seal without O-rings.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Thread Size Ace-Thred No. 25
Material PTFE