Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MICL/CLEC12A Antibody (812303R), DyLight 550, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2950RL
Description
MICL/CLEC12A Monoclonal antibody specifically detects MICL/CLEC12A in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
MICL/CLEC12A | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
160364 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
812303R | |
Flow Cytometry, CyTOF-ready | |
CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL | |
Mouse myeloma cell line NS0-derived recombinant mouse MICL/CLEC12A, Thr67-Arg267, Accession # Q504P2 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction