Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MICL/CLEC12A Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$149.33 - $394.26

Specifications

Antigen MICL/CLEC12A
Concentration 0.5 mg/mL
Dilution Western Blot 2-4ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227346SS
SDP
View Documents
Novus Biologicals
NBP227346SS
0.025 mg
Each for $149.33
Only null left
Add to Cart
 
NBP227346
SDP
View Documents
Novus Biologicals
NBP227346
0.1 mg
Each for $394.26
Only null left
Add to Cart
 
Description

Description

MICL/CLEC12A Polyclonal specifically detects MICL/CLEC12A in Human samples. It is validated for Western Blot.
Specifications

Specifications

MICL/CLEC12A
Western Blot 2-4ug/ml
Polyclonal
Purified
RUO
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL
CLEC12A
IgG
Protein A purified
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, 94% in chimpanzee, 61% in orangutan, white-cheeked gibbon, and rhesus monkey.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Human
Q5QGZ9
160364
A portion of amino acids170-220 from human CLEC12A was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.