Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Miconazole (Free Base), Fisher BioReagents
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 g
25 g
Packaging:
Amber Glass
Description
Miconazole is used as an antifungal agent.

Specifications
Specifications
Type | Miconazole |
Physical Form | Solid |
Melting Point | 135°C |
Ignition Residue | < 0.2% |
Loss on Drying | < 0.5% |
Packaging | Amber Glass |
Color | White |
Quantity | 1 g |
Assay Percent Range | 98% |
Merck Index | 15, 6257 |
Show More |
Safety and Handling

Hazard Statement | WARNING! |
Precautionary Statement | Emergency Overview Harmful if swallowed. May cause allergic respiratory reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting. NFPA Health:2 Flammability:0 Instability:0 |
Recommended Storage | RT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction