Learn More
World Precision Instrument Micro Cannula, 3", 0.4mm OD
SDP
Catalog No. 502538729
Encompass
This micro cannula is ideal for placement in the carotid or femoral artery of mice, rats, and other small animal blood vessels. It can be used with a pressure transducer (WPI’S BLPR2) for blood pressure measurement, or in conjunction with a micro-syringe injection system (like WPI’s UMPIII or MMP pumps). The incorporated standard female luer fitting makes connecting to existing experimental plumbing quick and easy. The cannula is provided with a contoured-tip stainless steel stylet (trocar) to facilitate placement using established techniques. A movable “shoulder” ring provides a tie-in point to prevent accidental removal. The cannula may be left in place for 2 hours or more, and with proper care and cleaning, may be re-used multiple times. Instructions are included.
Catalog No. 50-253-8729 Supplier World Precision Instrument Supplier No. KZ1101
This micro cannula is ideal for placement in the carotid or femoral artery of mice, rats, and other small animal blood vessels. It can be used with a pressure transducer (WPI’S BLPR2) for blood pressure measurement, or in conjunction with a micro-syringe injection system (like WPI’s UMPIII or MMP pumps). The incorporated standard female luer fitting makes connecting to existing experimental plumbing quick and easy. The cannula is provided with a contoured-tip stainless steel stylet (trocar) to facilitate placement using established techniques. A movable “shoulder” ring provides a tie-in point to prevent accidental removal. The cannula may be left in place for 2 hours or more, and with proper care and cleaning, may be re-used multiple times. Instructions are included.