Learn More
Stoelting™ Micro Clip
SDP
Stoelting™ Micro Clips provide an easy and secure method of stopping blood flow while performing procedures on vessels.
$197.20 - $229.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 7
Catalog Number Mfr. No. Model Size Price Quantity & Availability  
Catalog Number Mfr. No. Model Size Price Quantity & Availability  
10-001-250
SDP
View Documents
Stoelting™
5210665
Straight 0.75 x 4 mm
Each for $202.50
Only null left
Add to Cart
10-001-251
SDP
View Documents
Stoelting™
5210666
Straight 0.8 x 6 mm
Each for $197.20
Only null left
Add to Cart
10-001-252
SDP
View Documents
Stoelting™
5210667
Straight 1 x 8 mm
Each for $197.20
Only null left
Add to Cart
10-001-253
SDP
View Documents
Stoelting™
5210668
Straight 1.5 x 10 mm
Each for $197.20
Only null left
Add to Cart
10-001-254
SDP
View Documents
Stoelting™
5210669
Slight curve 1 x 6 mm
Each for $229.00
Only null left
Add to Cart
10-001-255
SDP
View Documents
Stoelting™
5210670
Strong curve 1 x 6 mm
Each for $229.00
Only null left
Add to Cart
10-001-256
SDP
View Documents
Stoelting™
5210671
45° Angle 1 x 6 mm
Each for $229.00
Only null left
Add to Cart
Description
Description
  • Using the Stoelting applying forceps, surgical clips can easily be positioned on the vessels.
  • Locking forceps allows for easy, precise, and secure grasping of the micro surgical clips.
  • Simply grasp the clip with the forceps to open the jaw, slid it over the vessels and release.
  • To remove surgical clips, simply perform the procedure in reverse.
  • Procedure does not compromise the vessels integrity.
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.